Sigma-Aldrich
Vanillic acid, ≥97%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Vanillic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
p-Anisic acid, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Pricing and availability is not currently available.
Supelco
Melting point standard 182-184°C, analytical standard
Pricing and availability is not currently available.
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Pricing and availability is not currently available.
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Ferulic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
3,4-Dihydroxybenzaldehyde, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methoxybenzoic acid, ReagentPlus®, 99%
Pricing and availability is not currently available.
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Pricing and availability is not currently available.