Merck
  • Leukotriene receptor antagonists enhance HCC treatment efficacy by inhibiting ADAMs and suppressing MICA shedding.
Leukotriene receptor antagonists enhance HCC treatment efficacy by inhibiting ADAMs and suppressing MICA shedding.
Cancer immunology, immunotherapy : CII (2020-07-20)
Jun Arai, Kaku Goto, Yumi Otoyama, Yoko Nakajima, Ikuya Sugiura, Atsushi Kajiwara, Masayuki Tojo, Yuki Ichikawa, Shojiro Uozumi, Yuu Shimozuma, Manabu Uchikoshi, Masashi Sakaki, Hisako Nozawa, Ryo Nakagawa, Ryosuke Muroyama, Naoya Kato, Hitoshi Yoshida
ABSTRACT
In our previous genome-wide association study, we demonstrated the association between MHC class I-related chain A (MICA) and hepatocellular carcinoma (HCC) development in patients with chronic hepatitis C. Increasing membrane-bound MICA (mMICA) in cancer cells by reducing MICA sheddases facilitates natural killer (NK) cell-mediated cytotoxicity. Our recent study clarified that A disintegrin and metalloproteases (ADAM), including ADAM9, are MICA sheddases in HCC, and that the suppression of ADAMs increases mMICA, demonstrating the rationality of mMICA-NK targeted therapy. Furthermore, we showed that regorafenib suppresses ADAM9 transcriptionally and translationally. A library of FDA-approved drugs was screened for more efficient inhibitors of ADAM9. Flow cytometry evaluation of the expression of mMICA after treatment with various candidate drugs identified leukotriene receptor antagonists as potential ADAM9 inhibitors. Furthermore, leukotriene receptor antagonists alone or in combination with regorafenib upregulated mMICA, which was in turn downregulated by leukotriene C4 and D4 via ADAM9 function. Our study demonstrates that leukotriene receptor antagonists could be developed as novel drugs for immunological control and suppression of ADAM9 in HCC. Further, leukotriene receptor antagonists should be explored as combination therapy partners with conventional multi-kinase inhibitors for developing therapeutic strategies with enhanced efficacies for HCC management and treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human ADAM9