Merck
  • Bexarotent Attenuated Chronic Constriction Injury-Induced Spinal Neuroinflammation and Neuropathic Pain by Targeting Mitogen-Activated Protein Kinase Phosphatase-1.
Bexarotent Attenuated Chronic Constriction Injury-Induced Spinal Neuroinflammation and Neuropathic Pain by Targeting Mitogen-Activated Protein Kinase Phosphatase-1.
The journal of pain (2019-01-21)
Yulong Gui, Shunyuan Duan, Lihong Xiao, Jing Tang, Aiyuan Li
ABSTRACT
It is widely accepted that neuroinflammation in the spinal cord contributes to the development of central sensitization in neuropathic pain. Mitogen-activated protein kinase (MAPK) activation plays a vital role in the development of neuroinflammation in the spinal cord. In this study, we investigated the effect of bexarotene (bex), a retinoid X receptor agonist, on MAPKs activation in chronic constriction injury (CCI)-induced neuropathic pain. The data showed that daily treatment with bex 50 mg/kg significantly alleviated CCI-induced nociceptive hypersensitivity in rats. Bex 50 mg/kg/day inhibited CCI-induced MAPKs (p38MAPK, ERK1/2, and JNK) activation and upregulation of proinflammatory factors (IL-1β, tumor necrosis factor-α and IL-6). Bex also reversed CCI-induced microglia activation in the ipsilateral spinal cord. Furthermore, bex treatment significantly upregulated MKP-1 in the spinal cord. These effects were completely abrogated by MKP-1 inhibitor BCI. These results indicated that bex relieved CCI-induced neuroinflammation and neuropathic pain by targeting MKP-1. Therefore, bex might be a potential agent for the treatment of neuropathic pain. PERSPECTIVE: Bex could relieve neuropathic pain behaviors in animals by reversing MKP-1 downregulation and MAPKs activation in the spinal cord. Therapeutic applications of bex may be extended beyond cutaneous T-cell lymphoma.
Sigma-Aldrich
(E/Z)-BCI hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.