Merck
  • The permeation of amphoteric drugs through artificial membranes--an in combo absorption model based on paracellular and transmembrane permeability.
The permeation of amphoteric drugs through artificial membranes--an in combo absorption model based on paracellular and transmembrane permeability.
Journal of medicinal chemistry (2009-12-02)
Kin Y Tam, Alex Avdeef, Oksana Tsinman, Na Sun
ABSTRACT
The permeability characteristics of 33 amphoteric drugs (about 64% zwitterions at physiological pH) were studied using the parallel artificial membrane permeability assay (PAMPA) at pH 6.5. The PAMPA data were modified to include the paracellular permeability component found in cellular monolayers based on a newly generalized version of a popular model devised for Caco-2 cells. These "in combo" PAMPA data were used to predict the human absolute bioavailability of the ampholytes. The analysis produced a good fit, with only five outliers whose transport properties, could be rationalized by (a) nonpassive permeability processes, (b) metabolic instability, and (c) the possible sensitivity to microclimate pH effects in the case of acidic ampholytes. With the exception of two compounds, all of the ampholytes with bioavailability <50% were predominantly transported by the paracellular route, surprisingly with several of the compounds having molecular weights exceeding 350 Da.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Sparfloxacin, ≥98.0% (HPLC)
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Melphalan, powder
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Supelco
Norfloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Famotidine
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)