Merck
  • Palmitoylation of the Bovine Foamy Virus Envelope Glycoprotein Is Required for Viral Replication.
Palmitoylation of the Bovine Foamy Virus Envelope Glycoprotein Is Required for Viral Replication.
Viruses (2020-12-31)
Keli Chai, Zhaohuan Wang, Yali Xu, Junshi Zhang, Juan Tan, Wentao Qiao
ABSTRACT
Membrane proteins of enveloped viruses have been reported to undergo palmitoylation, a post-translational modification often having a critical role in the function of these viral proteins and hence viral replication. In this study, we report that the foamy virus (FV) envelope (Env) glycoprotein is palmitoylated. Specifically, we found that bovine foamy virus (BFV) Env (BEnv) is palmitoylated at amino acid positions C58 and C59 by BDHHC3 and BDHHC20 in a DHHC motif-dependent manner. In addition, mutations C58S and C58/59S significantly decrease cell surface expression of BEnv, subviral particle (SVP) egress, and its membrane fusion activity, thus ultimately inhibiting BFV replication. The C59S mutation exerts a minor effect in this regard. Taken together, these data demonstrate that the function of BEnv in the context of BFV replication is under the regulation of palmitoylation.
Sigma-Aldrich
2-Bromohexadecanoic acid, ~97%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
Protein A Agarose, Protein A Agarose Beads for the purification of human, mouse & rabbit immunoglobins.
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Brij® O10