Merck
  • Inhibitory effects of polyphenols and their colonic metabolites on CYP2D6 enzyme using two different substrates.
Inhibitory effects of polyphenols and their colonic metabolites on CYP2D6 enzyme using two different substrates.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-09-18)
Eszter Fliszár-Nyúl, Violetta Mohos, Rita Csepregi, Přemysl Mladěnka, Miklós Poór
ABSTRACT
Polyphenolic compounds (including flavonoids, chalcones, phenolic acids, and furanocoumarins) represent a common part of our diet, but are also the active ingredients of several dietary supplements and/or medications. These compounds undergo extensive metabolism by human biotransformation enzymes and the microbial flora of the colon. CYP2D6 enzyme metabolizes approximately 25% of the drugs, some of which has narrow therapeutic window. Therefore, its inhibition can lead to the development of pharmacokinetic interactions and the disruption of drug therapy. In this study, the inhibitory effects of 17 plant-derived compounds and 19 colonic flavonoid metabolites on CYP2D6 were examined, employing two assays with different test substrates. The O-demethylation of dextromethorphan was tested employing CypExpress 2D6 kit coupled to HPLC analysis; while the O-demethylation of another CYP2D6 specific substrate (AMMC) was investigated in a plate reader assay with BioVision Fluorometric CYP2D6 kit. Interestingly, some compounds (e.g., bergamottin) inhibited both dextromethorphan and AMMC demethylation; however, certain substances proved to be inhibitors only in one of the assays applied. Our results demonstrate that some polyphenols and colonic metabolites are inhibitors of CYP2D6-catalyzed reactions. Nevertheless, the inhibitory effects showed strong substrate dependence.
Sigma-Aldrich
3-Hydroxy-4-methoxyphenylacetic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methylcatechol, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
3-(2,4-Dihydroxyphenyl)propionic acid, ≥95.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Pyroglutamic acid 2-naphthylamide, ≥99% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Hydroxymethyl)phenylacetic acid, 98%
Pricing and availability is not currently available.