Merck
  • Derivation of induced pluripotent stem cells (SDUKIi003-A) from a 20-year-old male patient diagnosed with Asperger syndrome.
Derivation of induced pluripotent stem cells (SDUKIi003-A) from a 20-year-old male patient diagnosed with Asperger syndrome.
Stem cell research (2020-09-12)
Morad Kamand, Mirolyuba Ilieva, Sheena Louise Forsberg, Mads Thomassen, Morten Meyer, Åsa Fex Svenningsen, Tanja Maria Michel
ABSTRACT
Autism spectrum disorder is a heterogenous neurodevelopmental disorder. The patients experience challenges in social interaction and communication skills as well as restricted and/or repetitive behaviors. To understand the molecular mechanisms underlying developmental brain disorders, patient-derived cellular models represent a useful tool. We have generated a human induced pluripotent stem cell line (SDUKIi003-A) from skin fibroblasts derived from a 20-year old male patient diagnosed with Asperger syndrome ("FYNEN-cohort" of Southern Denmark). The reprogramming of the fibroblasts was accomplished using integration-free episomal plasmids. Characterization validated the expression of pluripotency markers, differentiation into the three germ layers, and absence of chromosomal abnormalities.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
Trypsin-EDTA solution, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
Triton X-100, for molecular biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
€104.00
1 kg
Please contact Customer Service for Availability
€214.00
5 kg
Please contact Customer Service for Availability
€445.00