Merck
  • PasT of Escherichia coli sustains antibiotic tolerance and aerobic respiration as a bacterial homolog of mitochondrial Coq10.
PasT of Escherichia coli sustains antibiotic tolerance and aerobic respiration as a bacterial homolog of mitochondrial Coq10.
MicrobiologyOpen (2020-06-20)
Cinzia Fino, Martin Vestergaard, Hanne Ingmer, Fabien Pierrel, Kenn Gerdes, Alexander Harms
ABSTRACT
Antibiotic-tolerant persisters are often implicated in treatment failure of chronic and relapsing bacterial infections, but the underlying molecular mechanisms have remained elusive. Controversies revolve around the relative contribution of specific genetic switches called toxin-antitoxin (TA) modules and global modulation of cellular core functions such as slow growth. Previous studies on uropathogenic Escherichia coli observed impaired persister formation for mutants lacking the pasTI locus that had been proposed to encode a TA module. Here, we show that pasTI is not a TA module and that the supposed toxin PasT is instead the bacterial homolog of mitochondrial protein Coq10 that enables the functionality of the respiratory electron carrier ubiquinone as a "lipid chaperone." Consistently, pasTI mutants show pleiotropic phenotypes linked to defective electron transport such as decreased membrane potential and increased sensitivity to oxidative stress. We link impaired persister formation of pasTI mutants to a global distortion of cellular stress responses due to defective respiration. Remarkably, the ectopic expression of human coq10 largely complements the respiratory defects and decreased persister levels of pasTI mutants. Our work suggests that PasT/Coq10 has a central role in respiratory electron transport that is conserved from bacteria to humans and sustains bacterial tolerance to antibiotics.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxybenzoic acid-(phenyl-13C6), 99 atom % 13C
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Pricing and availability is not currently available.