Merck
  • A single-host fermentation process for the production of flavor lactones from non-hydroxylated fatty acids.
A single-host fermentation process for the production of flavor lactones from non-hydroxylated fatty acids.
Metabolic engineering (2019-08-14)
Eko Roy Marella, Jonathan Dahlin, Marie Inger Dam, Jolanda Ter Horst, Hanne Bjerre Christensen, Suresh Sudarsan, Guokun Wang, Carina Holkenbrink, Irina Borodina
ABSTRACT
Lactone flavors with fruity, milky, coconut, and other aromas are widely used in the food and fragrance industries. Lactones are produced by chemical synthesis or by biotransformation of plant-sourced hydroxy fatty acids. We established a novel method to produce flavor lactones from abundant non-hydroxylated fatty acids using yeast cell factories. Oleaginous yeast Yarrowia lipolytica was engineered to perform hydroxylation of fatty acids and chain-shortening via β-oxidation to preferentially twelve or ten carbons. The strains could produce γ-dodecalactone from oleic acid and δ-decalactone from linoleic acid. Through metabolic engineering, the titer was improved 4-fold, and the final strain produced 282 mg/L γ-dodecalactone in a fed-batch bioreactor. The study paves the way for the production of lactones by fermentation of abundant fatty feedstocks.
Sigma-Aldrich
Oleoyl coenzyme A lithium salt, ≥90% (HPLC)
Sigma-Aldrich
Palmitoyl coenzyme A lithium salt, ≥90%
Sigma-Aldrich
Decanoyl coenzyme A monohydrate, ≥90%
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Sigma-Aldrich
Myristoyl coenzyme A lithium salt, ≥80.0%
Sigma-Aldrich
Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Glass beads, acid-washed, 212-300 μm (50-70 U.S. sieve)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Methyl oleate, 99%
Sigma-Aldrich
Octanoyl coenzyme A lithium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Lauroyl coenzyme A lithium salt, ≥90% (HPLC)