Merck
  • Antisense oligonucleotide modulation of non-productive alternative splicing upregulates gene expression.
Antisense oligonucleotide modulation of non-productive alternative splicing upregulates gene expression.
Nature communications (2020-07-11)
Kian Huat Lim, Zhou Han, Hyun Yong Jeon, Jacob Kach, Enxuan Jing, Sebastien Weyn-Vanhentenryck, Mikaela Downs, Anna Corrionero, Raymond Oh, Juergen Scharner, Aditya Venkatesh, Sophina Ji, Gene Liau, Barry Ticho, Huw Nash, Isabel Aznarez
ABSTRACT
While most monogenic diseases are caused by loss or reduction of protein function, the need for technologies that can selectively increase levels of protein in native tissues remains. Here we demonstrate that antisense-mediated modulation of pre-mRNA splicing can increase endogenous expression of full-length protein by preventing naturally occurring non-productive alternative splicing and promoting generation of productive mRNA. Bioinformatics analysis of RNA sequencing data identifies non-productive splicing events in 7,757 protein-coding human genes, of which 1,246 are disease-associated. Antisense oligonucleotides targeting multiple types of non-productive splicing events lead to increases in productive mRNA and protein in a dose-dependent manner in vitro. Moreover, intracerebroventricular injection of two antisense oligonucleotides in wild-type mice leads to a dose-dependent increase in productive mRNA and protein in the brain. The targeting of natural non-productive alternative splicing to upregulate expression from wild-type or hypomorphic alleles provides a unique approach to treating genetic diseases.
Product Description
Millipore
Accutase cell detachment solution, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Sigma-Aldrich
Epidermal Growth Factor Protein, Human recombinant, The Epidermal Growth Factor (EGF) is a polypeptide growth factor which stimulates the proliferation of a wide range of epidermal & epithelial cells.