Merck
  • On-chip recapitulation of clinical bone marrow toxicities and patient-specific pathophysiology.
On-chip recapitulation of clinical bone marrow toxicities and patient-specific pathophysiology.
Nature biomedical engineering (2020-01-29)
David B Chou, Viktoras Frismantas, Yuka Milton, Rhiannon David, Petar Pop-Damkov, Douglas Ferguson, Alexander MacDonald, Özge Vargel Bölükbaşı, Cailin E Joyce, Liliana S Moreira Teixeira, Arianna Rech, Amanda Jiang, Elizabeth Calamari, Sasan Jalili-Firoozinezhad, Brooke A Furlong, Lucy R O'Sullivan, Carlos F Ng, Youngjae Choe, Susan Marquez, Kasiani C Myers, Olga K Weinberg, Robert P Hasserjian, Richard Novak, Oren Levy, Rachelle Prantil-Baun, Carl D Novina, Akiko Shimamura, Lorna Ewart, Donald E Ingber
ABSTRACT
The inaccessibility of living bone marrow (BM) hampers the study of its pathophysiology under myelotoxic stress induced by drugs, radiation or genetic mutations. Here, we show that a vascularized human BM-on-a-chip (BM chip) supports the differentiation and maturation of multiple blood cell lineages over 4 weeks while improving CD34+ cell maintenance, and that it recapitulates aspects of BM injury, including myeloerythroid toxicity after clinically relevant exposures to chemotherapeutic drugs and ionizing radiation, as well as BM recovery after drug-induced myelosuppression. The chip comprises a fluidic channel filled with a fibrin gel in which CD34+ cells and BM-derived stromal cells are co-cultured, a parallel channel lined by human vascular endothelium and perfused with culture medium, and a porous membrane separating the two channels. We also show that BM chips containing cells from patients with the rare genetic disorder Shwachman-Diamond syndrome reproduced key haematopoietic defects and led to the discovery of a neutrophil maturation abnormality. As an in vitro model of haematopoietic dysfunction, the BM chip may serve as a human-specific alternative to animal testing for the study of BM pathophysiology.
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Triton X-100