Merck
  • Golgi-associated microtubules are fast cargo tracks and required for persistent cell migration.
Golgi-associated microtubules are fast cargo tracks and required for persistent cell migration.
EMBO reports (2020-01-28)
Huiwen Hao, Jiahao Niu, Boxin Xue, Qian Peter Su, Menghan Liu, Junsheng Yang, Jinshan Qin, Shujuan Zhao, Congying Wu, Yujie Sun
ABSTRACT
Microtubules derived from the Golgi (Golgi MTs) have been implicated to play critical roles in persistent cell migration, but the underlying mechanisms remain elusive, partially due to the lack of direct observation of Golgi MT-dependent vesicular trafficking. Here, using super-resolution stochastic optical reconstruction microscopy (STORM), we discovered that post-Golgi cargos are more enriched on Golgi MTs and also surprisingly move much faster than on non-Golgi MTs. We found that, compared to non-Golgi MTs, Golgi MTs are morphologically more polarized toward the cell leading edge with significantly fewer inter-MT intersections. In addition, Golgi MTs are more stable and contain fewer lattice repair sites than non-Golgi MTs. Our STORM/live-cell imaging demonstrates that cargos frequently pause at the sites of both MT intersections and MT defects. Furthermore, by optogenetic maneuvering of cell direction, we demonstrate that Golgi MTs are essential for persistent cell migration but not for cells to change direction. Together, our study unveils the role of Golgi MTs in serving as a group of "fast tracks" for anterograde trafficking of post-Golgi cargos.
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO