Merck
  • Alterations of estrous cycle, 3β hydroxysteroid dehydrogenase activity and progesterone synthesis in female rats after exposure to hypobaric hypoxia.
Alterations of estrous cycle, 3β hydroxysteroid dehydrogenase activity and progesterone synthesis in female rats after exposure to hypobaric hypoxia.
Scientific reports (2020-02-28)
Snigdha Shaw, Utkarsha Kumar, Gopinath Bhaumik, M Prasanna Kumar Reddy, Bhuvnesh Kumar, Dishari Ghosh
ABSTRACT
The underlying mechanism regulating hypoxia induced alteration in female steroid hormones is first time explored in this study. To understand the mechanistic approach, female Sprague- Dawley rats were exposed to acute and chronic hypobaric hypoxia (282 mm-Hg, ~7620 m, 6 hours, 3 and 7 days). Estrous cycle, body weight, plasma progesterone and estradiol levels, morphology, histology and two key steroidogenic enzymes: 3ß hydroxysteroid dehydrogenase (HSD) and 17ß HSD activity of ovary and adrenal gland were studied. A persistent diestrous phase and a significant decrease in body weight were found in chronic hypoxia groups. Histological study suggested degenerative changes in ovarian corpus luteum of 7 days chronic hypobaric hypoxia (7CHH) group and a declined percentage of adrenocortical cells in 3 days chronic hypobaric hypoxia (3CHH) and 7CHH groups. Plasma estradiol level was unaltered, but progesterone level was decreased significantly in all hypoxic groups. Ovarian 3ß HSD activity was decreased significantly with increasing days of hypoxic treatment along with a significantly low adrenal 3ß HSD activity in 7CHH. In conclusion, hypobaric hypoxia causes a state of low circulatory progesterone level in females likely due to the degenerative changes in the female ovarian and adrenal tissues together with low steroidogenic 3ß HSD enzyme activity.
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Methyl Blue, powder or crystals
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate disodium salt, ≥97% (calc. based on dry substance, enzymatic)