Merck
  • Resveratrol protects against apoptosis induced by interleukin-1β in nucleus pulposus cells via activating mTOR/caspase-3 and GSK-3β/caspase-3 pathways.
Resveratrol protects against apoptosis induced by interleukin-1β in nucleus pulposus cells via activating mTOR/caspase-3 and GSK-3β/caspase-3 pathways.
Bioscience reports (2020-07-23)
Xiaohui Guo, Xiaoliang Bai, Feng Zhang, Long Zheng, Wenyuan Ding, Sidong Yang
ABSTRACT
The purpose of the present study was to investigate the specific downstream signaling pathway mediated by PI3K/Akt in resveratrol (RES) anti-apoptosis of nucleus pulposus cells (NPCs). Human NPCs were cultured and divided into six groups. Interleukin (IL)-1β was used to induce apoptosis and RES to inhibit apoptosis. Fluorescence-activated cell sorting (FACS) analysis was used to test apoptotic incidence of NPCs, cell counting kit-8 (CCK-8) assay was performed to detect cell viability, The expression level of caspase-3 mRNA was detected by RT-qPCR, and protein levels were determined by Western blot. Flow cytometry analysis showed that IL-1β increased the apoptosis rate of NPCs in each group, and RES significantly decreased the apoptosis rate, while rapamycin (RAPA) and SB216763 inhibited the effect of RES and increased the apoptosis rate again. Similarly, CCK-8 showed that IL-1β decreased activity of NPCs in each group, while RES increased cell activity, RAPA and SB216763 inhibited the effect of RES and decreased cell activity. RT-qPCR results showed IL-1β significantly increased the level of caspase-3 expression, but it was significantly decreased by using RES, RAPA and SB216763 respectively attenuated effects of RES. Western blot results showed that activated caspase-3 was inhibited by RES effect, and was up-regulated again after the addition of RAPA and SB216763. In addition, p-mTOR and p-GSK-3β were up-regulated by RES and down-regulated by RAPA and SB216763. RES can inhibit apoptosis induced by IL-1β in human NPCs. PI3K/Akt/mTOR/caspase-3 and PI3K/Akt/GSK-3β/caspase-3 pathways are potential mechanisms underlying this process.