Merck
  • Proteolytic enzymes in seminal plasma of domestic turkey (Meleagris gallopavo).
Proteolytic enzymes in seminal plasma of domestic turkey (Meleagris gallopavo).
Biology of reproduction (1993-02-01)
R J Thurston, N Korn, D P Froman, A B Bodine
ABSTRACT
Protease and basic amidase activity was found in the seminal plasma of the domestic turkey. Amidase activity, measured through use of N-alpha-benzoyl-DL-arginine-p-nitroanilide-HCL (BAPNA), was 23-28 times greater for turkey than for guinea fowl or chicken. Within the reproductive tract, seminal plasma from the vas deferens had much greater activity than testicular or epididymal fluids. Turkey seminal plasma enzyme (TSPE) purified by chromatography or isoelectric focusing showed three protein bands by PAGE, each resolving on SDS-PAGE into two subunits with molecular weights of approximately 28,000-32,000 and 38,000-44,000. One of the three proteins also contained a larger subunit (M(r) 76,000-81,000) thought to be transferrin. Turkey acrosin consisted of three subunits with molecular weights below 20,500. Acrosin, but not TSPE, was visualized in native gels with N-alpha-benzoyl-DL-arginine-beta-naphthylamide (BANA)/Fast Garnet stain. Michaelis constants (BAPNA) for TSPE, acrosin, and trypsin were 2.41 +/- 0.12 x 10(-4) M (n = 5), 4.96-6.03 x 10(-4) M (n = 2), and 6.76 +/- 0.95 x 10(-4) M (n = 6), respectively. TSPE, like acrosin and trypsin, was inhibited by benzamidine but not iodoacetamide. While all natural trypsin inhibitors tested inhibited acrosin, TSPE was not inhibited by ovomucoid from chicken or turkey egg white.
Sigma-Aldrich
Trypsin inhibitor from chicken egg white, Type III-O (free of ovoinhibitor)
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Millipore
Protein A Agarose, Protein A Agarose Beads for the purification of human, mouse & rabbit immunoglobins.