Merck
  • Cytotoxic activity and inhibitory effect on nitric oxide production of triterpene saponins from the roots of Physospermum verticillatum (Waldst & Kit) (Apiaceae).
Cytotoxic activity and inhibitory effect on nitric oxide production of triterpene saponins from the roots of Physospermum verticillatum (Waldst & Kit) (Apiaceae).
Bioorganic & medicinal chemistry (2009-05-27)
Rosa Tundis, Marco Bonesi, Brigitte Deguin, Monica R Loizzo, Federica Menichini, Filomena Conforti, François Tillequin, Francesco Menichini
ABSTRACT
Three triterpene saponins isolated from the roots of Physospermum verticillatum and identified as saikosaponin a (1), buddlejasaponin IV (2), and songarosaponin D (3) were investigated in vitro for their cytotoxic activity against a panel of seven different cancer cell lines including ACHN, C32, Caco-2, COR-L23, A375, A549, and Huh-7D12 cell lines. The hydrolysis of sugar unit was performed on saikosaponin a (1) to obtain saikosapogenin a (4). All isolated saponins exhibited strong cytotoxic activity against COR-L23 cell line with IC(50) values ranged from 0.4 to 0.6 microM. A similar activity was recorded for saikogenin a (4). None of the tested compounds affected the proliferation of skin fibroblasts 142BR suggesting a selective action against cancer cells. Moreover, buddlejasaponin IV (2) and songarosaponin D (3) exerted significant inhibition of NO production in LPS induced RAW 264.7 macrophages with IC(50) of 4.2 and 10.4 microM, respectively.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Vinblastine sulfate salt, ≥97% (HPLC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
COR-L23, 92031919