Merck
  • Identification of binding sites for ivacaftor on the cystic fibrosis transmembrane conductance regulator.
Identification of binding sites for ivacaftor on the cystic fibrosis transmembrane conductance regulator.
iScience (2021-06-19)
Onofrio Laselva, Zafar Qureshi, Zhi-Wei Zeng, Evgeniy V Petrotchenko, Mohabir Ramjeesingh, C Michael Hamilton, Ling-Jun Huan, Christoph H Borchers, Régis Pomès, Robert Young, Christine E Bear
ABSTRACT
Ivacaftor (VX-770) was the first cystic fibrosis transmembrane conductance regulator (CFTR) modulatory drug approved for the treatment of patients with cystic fibrosis. Electron cryomicroscopy (cryo-EM) studies of detergent-solubilized CFTR indicated that VX-770 bound to a site at the interface between solvent and a hinge region in the CFTR protein conferred by transmembrane (tm) helices: tm4, tm5, and tm8. We re-evaluated VX-770 binding to CFTR in biological membranes using photoactivatable VX-770 probes. One such probe covalently labeled CFTR at two sites as determined following trypsin digestion and analysis by tandem-mass spectrometry. One labeled peptide resides in the cytosolic loop 4 of CFTR and the other is located in tm8, proximal to the site identified by cryo-EM. Complementary data from functional and molecular dynamic simulation studies support a model, where VX-770 mediates potentiation via multiple sites in the CFTR protein.
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Anti-CFTR Antibody, clone MM13-4, clone MM13-4, Upstate®, from mouse