Merck
  • Tinagl1 Suppresses Triple-Negative Breast Cancer Progression and Metastasis by Simultaneously Inhibiting Integrin/FAK and EGFR Signaling.
Tinagl1 Suppresses Triple-Negative Breast Cancer Progression and Metastasis by Simultaneously Inhibiting Integrin/FAK and EGFR Signaling.
Cancer cell (2019-01-08)
Minhong Shen, Yi-Zhou Jiang, Yong Wei, Brian Ell, Xinlei Sheng, Mark Esposito, Jooeun Kang, Xiang Hang, Hanqiu Zheng, Michelle Rowicki, Lanjing Zhang, Weichung J Shih, Toni Celià-Terrassa, Yirong Liu, IIeana Cristea, Zhi-Ming Shao, Yibin Kang
ABSTRACT
Triple-negative breast cancer (TNBC) patients have the worst prognosis and distant metastasis-free survival among all major subtypes of breast cancer. The poor clinical outlook is further exacerbated by a lack of effective targeted therapies for TNBC. Here we show that ectopic expression and therapeutic delivery of the secreted protein Tubulointerstitial nephritis antigen-like 1 (Tinagl1) suppresses TNBC progression and metastasis through direct binding to integrin α5β1, αvβ1, and epidermal growth factor receptor (EGFR), and subsequent simultaneous inhibition of focal adhesion kinase (FAK) and EGFR signaling pathways. Moreover, Tinagl1 protein level is associated with good prognosis and reversely correlates with FAK and EGFR activation status in TNBC. Our results suggest Tinagl1 as a candidate therapeutic agent for TNBC by dual inhibition of integrin/FAK and EGFR signaling pathways.
Sigma-Aldrich
α Integrin Blocking and IHC Kit
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Sigma-Aldrich
β Integrin Screening Kit