Merck
  • Synthesis and SAR studies of 1,4-diazabicyclo[3.2.2]nonane phenyl carbamates--subtype selective, high affinity alpha7 nicotinic acetylcholine receptor agonists.
Synthesis and SAR studies of 1,4-diazabicyclo[3.2.2]nonane phenyl carbamates--subtype selective, high affinity alpha7 nicotinic acetylcholine receptor agonists.
Bioorganic & medicinal chemistry letters (2009-07-07)
Christopher J O'Donnell, Langu Peng, Brian T O'Neill, Eric P Arnold, Robert J Mather, Steven B Sands, Alka Shrikhande, Lorraine A Lebel, Douglas K Spracklin, Frank M Nedza
ABSTRACT
The synthesis and SAR studies about the bicyclic amine, carbamate linker and aromatic ring of a 1,4-diazabicyclo[3.2.2]nonane phenyl carbamate series of alpha7 nAChR agonists is described. The development of the medicinal chemistry strategy and SAR which led to the identification of 5 and 7aa as subtype selective, high affinity alpha7 agonists as excellent leads for further evaluation is discussed, along with key physicochemical and pharmacokinetic data highlighting their lead potential.
Product Description
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid