Merck
  • Staphylococcus aureus Panton-Valentine Leukocidin triggers an alternative NETosis process targeting mitochondria.
Staphylococcus aureus Panton-Valentine Leukocidin triggers an alternative NETosis process targeting mitochondria.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-11-27)
Viola Mazzoleni, Kiran Zimmermann, Anna Smirnova, Ivan Tarassov, Gilles Prévost
ABSTRACT
Panton-Valentine Leukocidin (PVL) is a bicomponent leukotoxin produced by 3%-10% of clinical Staphylococcus aureus (SA) strains involved in the severity of hospital and community-acquired infections. Although PVL was long known as a pore-forming toxin, recent studies have challenged the formation of a pore at the plasma membrane, while its endocytosis and the exact mode of action remain to be defined. In vitro immunolabeling of human neutrophils shows that Neutrophil Extracellular Traps (NETosis) is triggered by the action of purified PVL, but not by Gamma hemolysin CB (HlgCB), a structurally similar SA leukotoxin. PVL causes the ejection of chromatin fibers (NETs) decorated with antibacterial peptides independently of the NADPH oxidase oxidative burst. Leukotoxin partially colocalizes with mitochondria and enhances the production of reactive oxygen species from these organelles, while showing an increased autophagy, which results unnecessary for NETs ejection. PVL NETosis is elicited through Ca2+ -activated SK channels and Myeloperoxidase activity but is abolished by Allopurinol pretreatment of neutrophils. Moreover, massive citrullination of the histone H3 is performed by peptidyl arginine deiminases. Inhibition of this latter enzymes fails to abolish NET extrusion. Unexpectedly, PVL NETosis does not seem to involve Src kinases, which is the main kinase family activated downstream the binding of PVL F subunit to CD45 receptor, while the specific kinase pathway differs from the NADPH oxidase-dependent NETosis. PVL alone causes a different and specific form of NETosis that may rather represent a bacterial strategy conceived to disarm and disrupt the immune response, eventually allowing SA to spread.
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Anti-phospho-Erk1/2 (Thr202/Tyr204, Thr185/Tyr187)Antibody, recombinant clone AW39R, rabbit monoclonal, clone AW39R, Upstate®, from rabbit
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Hoechst 33258 solution, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining