Merck
  • Human physiomimetic model integrating microphysiological systems of the gut, liver, and brain for studies of neurodegenerative diseases.
Human physiomimetic model integrating microphysiological systems of the gut, liver, and brain for studies of neurodegenerative diseases.
Science advances (2021-01-31)
Martin Trapecar, Emile Wogram, Devon Svoboda, Catherine Communal, Attya Omer, Tenzin Lungjangwa, Pierre Sphabmixay, Jason Velazquez, Kirsten Schneider, Charles W Wright, Samuel Mildrum, Austin Hendricks, Stuart Levine, Julien Muffat, Meelim Jasmine Lee, Douglas A Lauffenburger, David Trumper, Rudolf Jaenisch, Linda G Griffith
ABSTRACT
Slow progress in the fight against neurodegenerative diseases (NDs) motivates an urgent need for highly controlled in vitro systems to investigate organ-organ- and organ-immune-specific interactions relevant for disease pathophysiology. Of particular interest is the gut/microbiome-liver-brain axis for parsing out how genetic and environmental factors contribute to NDs. We have developed a mesofluidic platform technology to study gut-liver-cerebral interactions in the context of Parkinson's disease (PD). It connects microphysiological systems (MPSs) of the primary human gut and liver with a human induced pluripotent stem cell-derived cerebral MPS in a systemically circulated common culture medium containing CD4+ regulatory T and T helper 17 cells. We demonstrate this approach using a patient-derived cerebral MPS carrying the PD-causing A53T mutation, gaining two important findings: (i) that systemic interaction enhances features of in vivo-like behavior of cerebral MPSs, and (ii) that microbiome-associated short-chain fatty acids increase expression of pathology-associated pathways in PD.
Sigma-Aldrich
[Leu15]-Gastrin I human, ≥95% (HPLC)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin solution, 30% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, Molecular Biology