Merck
  • A novel long intergenic noncoding RNA indispensable for the cleavage of mouse two-cell embryos.
A novel long intergenic noncoding RNA indispensable for the cleavage of mouse two-cell embryos.
EMBO reports (2016-08-09)
Jiaqiang Wang, Xin Li, Leyun Wang, Jingyu Li, Yanhua Zhao, Gerelchimeg Bou, Yufei Li, Guanyi Jiao, Xinghui Shen, Renyue Wei, Shichao Liu, Bingteng Xie, Lei Lei, Wei Li, Qi Zhou, Zhonghua Liu
ABSTRACT
Endogenous retroviruses (ERVs) are transcriptionally active in cleavage stage embryos, yet their functions are unknown. ERV sequences are present in the majority of long intergenic noncoding RNAs (lincRNAs) in mouse and humans, playing key roles in many cellular processes and diseases. Here, we identify LincGET as a nuclear lincRNA that is GLN-, MERVL-, and ERVK-associated and essential for mouse embryonic development beyond the two-cell stage. LincGET is expressed in late two- to four-cell mouse embryos. Its depletion leads to developmental arrest at the late G2 phase of the two-cell stage and to MAPK signaling pathway inhibition. LincGET forms an RNA-protein complex with hnRNP U, FUBP1, and ILF2, promoting the cis-regulatory activity of long terminal repeats (LTRs) in GLN, MERVL, and ERVK (GLKLTRs), and inhibiting RNA alternative splicing, partially by downregulating hnRNP U, FUBP1, and ILF2 protein levels. Hnrnpu or Ilf2 mRNA injection at the pronuclear stage also decreases the preimplantation developmental rate, and Fubp1 mRNA injection at the pronuclear stage causes a block at the two-cell stage. Thus, as the first functional ERV-associated lincRNA, LincGET provides clues for ERV functions in cleavage stage embryonic development.
Product Description
Roche
DIG DNA Labeling and Detection Kit