Merck
  • Synthesis and biological evaluation of new glutamic acid-based inhibitors of MurD ligase.
Synthesis and biological evaluation of new glutamic acid-based inhibitors of MurD ligase.
Bioorganic & medicinal chemistry letters (2008-11-19)
Tihomir Tomasić, Nace Zidar, Veronika Rupnik, Andreja Kovac, Didier Blanot, Stanislav Gobec, Danijel Kikelj, Lucija Peterlin Masic
ABSTRACT
Mur ligases catalyze the biosynthesis of the UDP-MurNAc-pentapeptide precursor of peptidoglycan, an essential polymer of bacterial cell-wall. They constitute attractive targets for the development of novel antibacterial agents. Here we report on the synthesis of a series of 2,4-diaminoquinazolines, quinazoline-2,4(1H,3H)-diones, 5-benzylidenerhodanines and 5-benzylidenethiazolidine-2,4-diones and their inhibitory activities against MurD from Escherichia coli. Compounds (R)-27 and (S)-27 showed inhibitory activity against MurD with IC(50) values of 174 and 206 microM, respectively, which makes them promising starting points for optimization.
Sigma-Aldrich
Folic acid, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Folic acid, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Pricing and availability is not currently available.