Merck
  • Propagation of goat putative spermatogonial stem cells under growth factors defined serum-free culture conditions.
Propagation of goat putative spermatogonial stem cells under growth factors defined serum-free culture conditions.
Cytotechnology (2020-03-04)
Ankur Sharma, Syed Mohmad Shah, Manish Tiwari, Mayank Roshan, Manoj Kumar Singh, Suresh Kumar Singla, Prabhat Palta, Radhay Sham Manik, Manmohan Singh Chauhan
ABSTRACT
In the present study, we used a serum-free culture media to propagate goat putative spermatogonial stem cells (SSCs) and evaluated the effect of crucial growth factors on relative expression of some SSC markers and self-renewal related genes. The enriched SSCs were cultured on a homologous Sertoli cell feeder layer in KO-DMEM supplemented with 10% KOSR. Putative SSC colonies emerged between day 6 and 10 which were then characterized by the expression of numerous spermatogonial and pluripotency related markers. After 15 days of subculture, the relative mRNA expression study revealed that 40 ng/mL concentration of Glial cell line-derived neurotrophic factor (GDNF) upregulated the expression of BCL6B, ID4, PLZF, and UCHL1. Moreover, the supplementation of GDNF + bFGF up-regulated the expression of PLZF and BCL6B. UCHL1 expression was higher after addition of GDNF + LIF while, THY1 overexpressed in response to the addition of GDNF + CSF1. These results demonstrated that the goat SSCs were efficiently propagated using a KOSR based serum-free media and the growth factor supplementation markedly influences their gene expression profile.