Merck
  • Lysine-specific demethylase-1 (LSD1) depletion disrupts monogenic and monoallelic odorant receptor (OR) expression in an olfactory neuronal cell line.
Lysine-specific demethylase-1 (LSD1) depletion disrupts monogenic and monoallelic odorant receptor (OR) expression in an olfactory neuronal cell line.
Molecular and cellular neurosciences (2017-04-18)
Rutesh N Vyas, Diane Meredith, Robert P Lane
ABSTRACT
Function of the mammalian olfactory system depends on specialized olfactory sensory neurons (OSNs) that each express only one allele ("monoallelic") of one odorant receptor (OR) gene ("monogenic"). The lysine-specific demethylase-1 (LSD1) protein removes activating H3K4 or silencing H3K9 methylation marks in a variety of developmental contexts, and is thought to be important for proper OR regulation. Most of the focus in the field has been on a potential "activating" function for LSD1; e.g., in the demethylation of H3K9 associated with the expressed OR allele. Here we show that depletion of LSD1 in an immortalized olfactory-placode-derived cell line (OP6) results in multigenic and multiallelic OR transcription per cell, while not seemingly disrupting the ability of these cells to activate new OR genes during clonal expansion. These results are consistent with LSD1 having a role in silencing additional OR alleles, as opposed to being required for the activation of OR alleles, within the OP6 cellular context.
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Co-Rest/RCOR1 Antibody, clone K72/8, clone K72/8, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315, rabbit monoclonal, culture supernatant, clone MC315, Upstate®