Merck
  • Elevated circulating endothelial cell-derived microparticle levels in patients with liver cirrhosis: a preliminary report.
Elevated circulating endothelial cell-derived microparticle levels in patients with liver cirrhosis: a preliminary report.
Clinical and experimental hepatology (2015-11-01)
Jolanta Zuwała-Jagiełło, Krzysztof Adam Simon, Monika Pazgan-Simon
ABSTRACT
To determine plausible associations between liver cirrhosis and circulating endothelial cell-derived microparticles (EMPs), vascular endothelial growth factor (VEGF) levels and plasma nitric oxide (NO) metabolites. Sixty patients with cirrhosis and 20 healthy control subjects were enrolled in the study. Circulating EMPs from platelet-poor plasma samples were examined by flow cytometry. These microparticles were categorized into endothelial cell-derived activated MPs (EMP-ac) (CD31+ CD42b- AN-V-) and endothelial cell-derived apoptotic MPs (EMP-ap) (CD31+ CD42b- AN-V+). Plasma VEGF levels were measured by enzyme-linked immunosorbent assay. Plasma NO metabolites (NOx-) levels were determined using a Greiss reaction method. Compared with the healthy control subjects, the patients with cirrhosis showed a significant increase in plasma levels of both phenotypes of EMPs. When the presence of ascites was considered, the plasma levels of EMP-ap were higher (p < 0.01), as well as NOx- (p < 0.05). EMP-ap positively correlated with VEGF level in all cirrhotic patients and this correlation was stronger in decompensated cirrhotic patients. In multivariate logistic regression analysis, the independent factors associated with the presence of ascites were high EMP-ap levels and elevated VEGF levels. Elevated plasma levels of EMP-ap in addition to high levels of VEGF might be considered as valuable parameters for predicting the occurrence of ascites in cirrhotic patients.
Sigma-Aldrich
Nitrite/Nitrate Assay Kit, colorimetric, used for detection of nitric oxide metabolites