Merck
  • Identification of small molecules rescuing fragile X syndrome phenotypes in Drosophila.
Identification of small molecules rescuing fragile X syndrome phenotypes in Drosophila.
Nature chemical biology (2008-03-11)
Shuang Chang, Steven M Bray, Zigang Li, Daniela C Zarnescu, Chuan He, Peng Jin, Stephen T Warren
ABSTRACT
Fragile X syndrome is caused by the functional loss of the fragile X mental retardation 1 (FMR1) gene. Deletion of the FMR1 ortholog in Drosophila melanogaster (Fmr1) recapitulates many phenotypes associated with fragile X syndrome. We have discovered that Fmr1 mutant Drosophila die during development when reared on food containing increased levels of glutamate, which is consistent with the theory that FMR1 loss results in excess glutamate signaling. Using this lethal phenotype, we screened a chemical library of 2,000 compounds and identified nine molecules that rescued the lethality, including three that implicate the GABAergic inhibitory pathway. Indeed, GABA treatment rescued several known Fmr1 mutant phenotypes in flies, including mushroom bodies defects, excess Futsch translation and abnormal male courtship behavior. These data are consistent with GABAergic inhibition of the enhanced excitatory pathway in fragile X syndrome. In addition, our screen reveals that the muscarinic cholinergic receptors may have a role in fragile X syndrome in parallel to the GABAergic pathway. These results point to potential therapeutic approaches for treating fragile X syndrome.
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Pilocarpine nitrate salt, ≥98% (HPLC)
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, 99%
Sigma-Aldrich
2-Aminoacetophenone hydrochloride, 99%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
Pilocarpine nitrate, meets USP testing specifications
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS