Merck
  • Selenium supplementation inhibits IGF-1 signaling and confers methionine restriction-like healthspan benefits to mice.
Selenium supplementation inhibits IGF-1 signaling and confers methionine restriction-like healthspan benefits to mice.
eLife (2021-03-31)
Jason D Plummer, Spike Dl Postnikoff, Jessica K Tyler, Jay E Johnson
ABSTRACT
Methionine restriction (MR) dramatically extends the healthspan of several organisms. Methionine-restricted rodents have less age-related pathology and increased longevity as compared with controls, and recent studies suggest that humans might benefit similarly. Mechanistically, it is likely that the decreased IGF-1 signaling that results from MR underlies the benefits of this regimen. Thus, we hypothesized that interventions that decrease IGF-1 signaling would also produce MR-like healthspan benefits. Selenium supplementation inhibits IGF-1 signaling in rats and has been studied for its putative healthspan benefits. Indeed, we show that feeding mice a diet supplemented with sodium selenite results in an MR-like phenotype, marked by protection against diet-induced obesity, as well as altered plasma levels of IGF-1, FGF-21, adiponectin, and leptin. Selenomethionine supplementation results in a similar, albeit less robust response, and also extends budding yeast lifespan. Our results indicate that selenium supplementation is sufficient to produce MR-like healthspan benefits for yeast and mammals.
Sigma-Aldrich
Histone Deacetylase Assay Kit, Fluorometric, sufficient for 100 assays (96 well plates)
Sigma-Aldrich
Alkaline Phosphatase Detection Kit, Fluorescence
Sigma-Aldrich
AST Activity Assay Kit, sufficient for 100 colorimetric tests
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.