Merck
  • lncRNA CEBPA-AS1 Overexpression Inhibits Proliferation and Migration and Stimulates Apoptosis of OS Cells via Notch Signaling.
lncRNA CEBPA-AS1 Overexpression Inhibits Proliferation and Migration and Stimulates Apoptosis of OS Cells via Notch Signaling.
Molecular therapy. Nucleic acids (2020-03-13)
Peng Xia, Rui Gu, Wei Zhang, Yi-Fu Sun
ABSTRACT
Osteosarcoma (OS) is the most common primary bone malignancy derived from primitive bone-forming mesenchymal cells. Long noncoding RNA (lncRNA) expression profiles have been intensively studied for their involvement in OS. Herein, we clarify whether lncRNA CEBPA-AS1 is a regulator of NCOR2 in OS cells. Microarray-based expression analysis identified OS-related differentially expressed lncRNA and predicted microRNAs (miRs) binding to lncRNA and mRNA. lncRNA CEBPA-AS1 and NCOR2 were found to be weakly expressed in OS tissues and cells. Next, functional investigation revealed that lncRNAs CEBPA-AS1 bound to miR-10b-5p to upregulate NCOR2. Following that, gene-targeted knockdown and overexpressed recombinant vectors of lncRNA CEBPA-AS1 and NCOR2 were constructed to explore the effects of lncRNA CEBPA-AS1 and NCOR2 on cell proliferation, differentiation, migration, and apoptosis. Finally, tumor formation was measured in nude mice. lncRNA CEBPA-AS1 overexpression or NCOR2 elevation inhibited cell proliferation and migration, and alkaline phosphatase (ALP) and bone gla protein (BGP) activity, while enhancing apoptosis and tumor formation. Furthermore, NCOR2 was elevated in response to lncRNA CEBPA-AS1 overexpression, thus repressing the Notch signaling pathway. Taken together, lncRNA CEBPA-AS1 overexpression inhibits OS progression through diminishing activation of the Notch signaling pathway via upregulating NCOR2. Therefore, lncRNA CEBPA-AS1 may serve as a molecular target for treating OS.
Product Description
Roche
Biotin RNA Labeling Mix, sufficient for 20 reactions (transcription), pkg of 40 μL, solution
Pricing and availability is not currently available.