Merck
  • Diagnosis, pathogenesis, treatment, and prognosis of hereditary fibrinogen A alpha-chain amyloidosis.
Diagnosis, pathogenesis, treatment, and prognosis of hereditary fibrinogen A alpha-chain amyloidosis.
Journal of the American Society of Nephrology : JASN (2008-12-17)
Julian D Gillmore, Helen J Lachmann, Dorota Rowczenio, Janet A Gilbertson, Cai-Hong Zeng, Zhi-Hong Liu, Lei-Shi Li, Ashutosh Wechalekar, Philip N Hawkins
ABSTRACT
Mutations in the fibrinogen A alpha-chain gene are the most common cause of hereditary renal amyloidosis in the United Kingdom. Previous reports of fibrinogen A alpha-chain amyloidosis have been in isolated kindreds, usually in the context of a novel amyloidogenic mutation. Here, we describe 71 patients with fibrinogen amyloidosis, who were prospectively studied at the UK National Amyloidosis Centre. Median age at presentation was 58 yr, and renal involvement led to diagnosis in all cases. Even after a median follow-up of 4 yr, clinically significant extra-renal disease was rare. Renal histology was characteristic: striking glomerular enlargement with almost complete obliteration of the normal architecture by amyloid deposition and little or no vascular or interstitial amyloid. We discovered four amyloidogenic mutations in fibrinogen (P552H, E540V, T538K, and T525fs). A family history of renal disease was frequently absent. Median time from presentation to ESRD was 4.6 yr, and the estimated median patient survival from presentation was 15.2 yr. Among 44 patients who reached ESRD, median survival was 9.3 yr. Twelve renal transplants survived for a median of 6.0 (0-12.2) yr. Seven grafts had failed after median follow up from transplantation of 5.8 yr, including three from recurrent amyloid after 5.8, 6.0, and 7.4 yr; three grafts failed immediately for surgical reasons and one failed from transplant glomerulopathy after 5.8 yr with no histological evidence of amyloid. At censor, the longest surviving graft was 12.2 yr. In summary, fibrinogen amyloidosis is predominantly a renal disease characterized by variable penetrance, distinctive histological appearance, proteinuria, and progressive renal impairment. Survival is markedly better than observed with systemic AL amyloidosis, and outcomes with renal replacement therapy are comparable to those for age-matched individuals with nondiabetic renal disease.
Sigma-Aldrich
Anti-Hsp90α Mouse mAb (EMD-17D7), liquid, clone EMD-17D7, Calbiochem®