Merck
  • Consumption of Quercetin and Quercetin-Containing Apple and Cherry Extracts Affects Blood Glucose Concentration, Hepatic Metabolism, and Gene Expression Patterns in Obese C57BL/6J High Fat-Fed Mice.
Consumption of Quercetin and Quercetin-Containing Apple and Cherry Extracts Affects Blood Glucose Concentration, Hepatic Metabolism, and Gene Expression Patterns in Obese C57BL/6J High Fat-Fed Mice.
The Journal of nutrition (2016-04-08)
Sarah M Snyder, Bingxin Zhao, Ting Luo, Clive Kaiser, George Cavender, Jill Hamilton-Reeves, Debra K Sullivan, Neil F Shay
ABSTRACT
Intake of polyphenols and polyphenol-rich fruit extracts has been shown to reduce markers of inflammation, diabetes, and hepatic complications that result from the consumption of a high-fat (HF) diet. The objective of this study was to determine whether mice fed polyphenol-rich apple peel extract (AE), cherry extract (CE), and quercetin, a phytochemical abundant in fruits including apples and cherries, would modulate the harmful effects of adiposity on blood glucose regulation, endocrine concentrations, and hepatic metabolism in HF-fed C57BL/6J male mice. Groups of 8-wk-old mice (n = 8 each) were fed 5 diets for 10 wk, including low-fat (LF; 10% of total energy) and HF (60% of total energy) control diets and 3 HF diets containing polyphenol-rich AE, CE, and quercetin (0.2% wt:wt). Also, an in vitro study used HepG2 cells exposed to quercetin (0-100 μmol/L) to determine whether intracellular lipid accumulation could be modulated by this phytochemical. Mice fed the HF control diet consumed 36% more energy, gained 14 g more body weight, and had ∼50% elevated blood glucose concentrations (all P < 0.05) than did LF-fed mice. Mice fed HF diets containing AE, CE, or quercetin became as obese as HF-fed mice, but had significantly lower blood glucose concentrations after food deprivation (-36%, -22%, -22%, respectively; P < 0.05). Concentrations of serum C-reactive protein were reduced 29% in quercetin-fed mice compared with HF-fed controls (P < 0.05). A qualitative evaluation of liver tissue sections suggested that fruit phytochemicals may reduce hepatic lipid accumulation. A quantitative analysis of lipid accumulation in HepG2 cells demonstrated a dose-dependent decrease in lipid content in cells treated with 0-100 μmol quercetin/L (P < 0.05). In mice, consumption of AE, CE, or quercetin appears to modulate some of the harmful effects associated with the consumption of an obesogenic HF diet. Furthermore, in a cell culture model, quercetin was shown to reduce intracellular lipid accumulation in a dose-dependent fashion.
Product Description
Millipore
MILLIPLEX® Mouse Adipokine Magnetic Bead Panel - Endocrine Multiplex Assay, The analytes available for this multiplex kit are: IL-6, Insulin, Leptin, MCP-1, PAI-1 (Total), Resistin, TNF-α.
Millipore
MILLIPLEX® Mouse Acute Phase Magnetic Bead Panel 2 - Cardiovascular Disease Multiplex Assay, The analytes available for this multiplex kit are: Adipsin, Alpha-1 Acid Glycoprotein (AGP), Alpha-2 Macroglobulin (A2M), C-Reactive Protein, Haptoglobin, SAP.