Merck
  • Ribosome-Targeting Antibiotics Impair T Cell Effector Function and Ameliorate Autoimmunity by Blocking Mitochondrial Protein Synthesis.
Ribosome-Targeting Antibiotics Impair T Cell Effector Function and Ameliorate Autoimmunity by Blocking Mitochondrial Protein Synthesis.
Immunity (2020-11-26)
Luís Almeida, Ayesha Dhillon-LaBrooy, Carla N Castro, Nigatu Adossa, Guilhermina M Carriche, Melanie Guderian, Saskia Lippens, Sven Dennerlein, Christina Hesse, Bart N Lambrecht, Luciana Berod, Leif Schauser, Bruce R Blazar, Markus Kalesse, Rolf Müller, Luís F Moita, Tim Sparwasser
ABSTRACT
While antibiotics are intended to specifically target bacteria, most are known to affect host cell physiology. In addition, some antibiotic classes are reported as immunosuppressive for reasons that remain unclear. Here, we show that Linezolid, a ribosomal-targeting antibiotic (RAbo), effectively blocked the course of a T cell-mediated autoimmune disease. Linezolid and other RAbos were strong inhibitors of T helper-17 cell effector function in vitro, showing that this effect was independent of their antibiotic activity. Perturbing mitochondrial translation in differentiating T cells, either with RAbos or through the inhibition of mitochondrial elongation factor G1 (mEF-G1) progressively compromised the integrity of the electron transport chain. Ultimately, this led to deficient oxidative phosphorylation, diminishing nicotinamide adenine dinucleotide concentrations and impairing cytokine production in differentiating T cells. In accordance, mice lacking mEF-G1 in T cells were protected from experimental autoimmune encephalomyelitis, demonstrating that this pathway is crucial in maintaining T cell function and pathogenicity.
Sigma-Aldrich
Chaetocin from Chaetomium minutum, ≥95% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
DIF-1, ≥98% (HPLC)
Sigma-Aldrich
Tigecycline hydrate, ≥98% (HPLC)
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Sodium ethoxide, technical, ≥95% (T)
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile