Merck
  • Reduced proteasome activity in the aging brain results in ribosome stoichiometry loss and aggregation.
Reduced proteasome activity in the aging brain results in ribosome stoichiometry loss and aggregation.
Molecular systems biology (2020-06-20)
Erika Kelmer Sacramento, Joanna M Kirkpatrick, Mariateresa Mazzetto, Mario Baumgart, Aleksandar Bartolome, Simone Di Sanzo, Cinzia Caterino, Michele Sanguanini, Nikoletta Papaevgeniou, Maria Lefaki, Dorothee Childs, Sara Bagnoli, Eva Terzibasi Tozzini, Domenico Di Fraia, Natalie Romanov, Peter H Sudmant, Wolfgang Huber, Niki Chondrogianni, Michele Vendruscolo, Alessandro Cellerino, Alessandro Ori
ABSTRACT
A progressive loss of protein homeostasis is characteristic of aging and a driver of neurodegeneration. To investigate this process quantitatively, we characterized proteome dynamics during brain aging in the short-lived vertebrate Nothobranchius furzeri combining transcriptomics and proteomics. We detected a progressive reduction in the correlation between protein and mRNA, mainly due to post-transcriptional mechanisms that account for over 40% of the age-regulated proteins. These changes cause a progressive loss of stoichiometry in several protein complexes, including ribosomes, which show impaired assembly/disassembly and are enriched in protein aggregates in old brains. Mechanistically, we show that reduction of proteasome activity is an early event during brain aging and is sufficient to induce proteomic signatures of aging and loss of stoichiometry in vivo. Using longitudinal transcriptomic data, we show that the magnitude of early life decline in proteasome levels is a major risk factor for mortality. Our work defines causative events in the aging process that can be targeted to prevent loss of protein homeostasis and delay the onset of age-related neurodegeneration.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Marizomib, ≥95% (HPLC), (Salinospora tropica)