Merck
  • Electronic effects of para-substitution on acetophenones in the reaction of rat liver 3alpha-hydroxysteroid dehydrogenase.
Electronic effects of para-substitution on acetophenones in the reaction of rat liver 3alpha-hydroxysteroid dehydrogenase.
Bioorganic & medicinal chemistry (2007-11-17)
Koji Uwai, Noboru Konno, Yuka Yasuta, Mitsuhiro Takeshita
ABSTRACT
Stereoselective reductive metabolism of various p-substituted acetophenone derivatives was studied using isolated rat liver 3alpha-hydroxysteroid dehydrogenase (3alpha-HSD). Kinetic experiments were performed and analyzed by measuring the products by HPLC using a chiral column. The results demonstrated that the presence of an electron-withdrawing substituent on the benzene ring plays an important role in determining the reduction rate in the syntheses of various (S)-alcohols from their corresponding carbonyl compounds. A plot of log {(V(max)/K(m))X/(V(max)/K(m))H} versus the substituent parameter (pi, sigma(para), Es) shows an increasing rate mainly for electron-withdrawing substituents, with a correlation coefficient (r(2)) of 0.97 which was obtained for triplicate data that were significant at the p<0.0001 level. With this in mind, new drugs can be designed that exploit this reduction pathway by introducing an electron-withdrawing group adjacent to the reduction site when a reduction reaction is desired, or by adding an electron-donating group when minimization of the reduction pathway is desired.
Sigma-Aldrich
Acetanisole, ≥98%, FCC, FG
Sigma-Aldrich
4′-Methylacetophenone, 95%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
4′-Methylacetophenone, ≥95%, FCC, FG
Sigma-Aldrich
4-Acetylbenzonitrile, 99%
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
4′-Bromoacetophenone, 98%
Sigma-Aldrich
4′-Methoxyacetophenone, 99%
Sigma-Aldrich
3α-Hydroxysteroid Dehydrogenase from Pseudomonas testosteroni, ≥15 units/mg protein