Merck
  • Folic acid supplementation acts as a chemopreventive factor in tumorigenesis of hepatocellular carcinoma by inducing H3K9Me2-dependent transcriptional repression of LCN2.
Folic acid supplementation acts as a chemopreventive factor in tumorigenesis of hepatocellular carcinoma by inducing H3K9Me2-dependent transcriptional repression of LCN2.
Oncotarget (2021-03-05)
Yin-Ling Zhang, Geng Xue, Hui Miao, Chuan-Chuan Zhou, Shu-Han Sun, Yi Zhang
ABSTRACT
The effects and mechanisms of folic acid (FA) as a chemopreventive agent for tumorigenesis of hepatocellular carcinoma (HCC) remain unclear. In this study, the QSG-7701, a human normal liver cell line, was cultured in different FA levels (High, Normal or No) for 6 months. Then, the biological characteristics, the expression of main stem cell-like genes or epithelial-mesenchymal transition (EMT) related genes and the tumorigenicity in vivo of cells cultured in different treatment groups were detected. Our results showed that No FA improved the malignant transformation of cells but High FA depressed the malignant transformation. Meanwhile, cells in different treatment groups were mapped by transcriptome sequencing. Then the relativity of increased LCN2 and decreased FA level was identified and confirmed in vitro and vivo. We also revealed that intracellular control of LCN2 would recover the effects of FA on cell proliferation, cell cycle and tumor formation in vitro and vivo. Finally, our studies displayed that increased FA level induced the down-regulation of LCN2 not by DNA hypermethylation of LCN2 promoter but by promoting the level of histone H3 lysine 9 di-methylation (H3K9Me2) in LCN2 promoter. In conclusion, our studies disclosed the chemopreventive effect of FA supplementation on hepatocarcinogenesis, which partial attributed to the inhibition of LCN2 by regulating histone methylation in promoter. Our results provide a potential mechanism of the chemoprevention of FA supplementation on tumorigenesis of HCC and may be helpful in developing treatment target against HCC.
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Immunoglobulin G from human serum, ≥95% (GE)
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), colorimetric, The CHEMICON Cell Invasion Assay Kit uses a 24-well plate, with 8 um pores, which provides an efficient system for evaluating the invasion of tumor cells through a basement membrane model.