Merck
  • Polymeric nanoparticles and microparticles for the delivery of peptides, biologics, and soluble therapeutics.
Polymeric nanoparticles and microparticles for the delivery of peptides, biologics, and soluble therapeutics.
Journal of controlled release : official journal of the Controlled Release Society (2015-09-12)
Robert F Pagels, Robert K Prud'homme
ABSTRACT
Biologically derived therapeutics, or biologics, are the most rapidly growing segment of the pharmaceutical marketplace. However, there are still unmet needs in improving the delivery of biologics. Injectable polymeric nanoparticles and microparticles capable of releasing proteins and peptides over time periods as long as weeks or months have been a major focus in the effort to decrease the frequency of administration. These particle systems fit broadly into two categories: those composed of hydrophilic and those composed of hydrophobic polymeric scaffolds. Here we review the factors that contribute to the slow and controlled release from each class of particle, as well as the effects of synthesis parameters and product design on the loading, encapsulation efficiency, biologic integrity, and release profile. Generally, hydrophilic scaffolds are ideal for large proteins while hydrophobic scaffolds are more appropriate for smaller biologics without secondary structure. Here we also introduce a Flash NanoPrecipitation method that has been adopted for encapsulating biologics in nanoparticles (40-200nm) at high loadings (50-75wt.%) and high encapsulation efficiencies. The hydrophilic gel interior and hydrophobic shell provide an opportunity to combine the best of both classes of injectable polymeric depots.
Sigma-Aldrich
NanoFabTx NanoFlash PEG-PLA drug formulation screening kit, for CIJ synthesis of nanoparticles
Sigma-Aldrich
NanoFabTx PEG-PLA drug formulation screening kit, for synthesis of PEGylated nanoparticles
Sigma-Aldrich
NanoFabTx NanoFlash PEG-PCL drug formulation screening kit, for CIJ synthesis of nanoparticles
Sigma-Aldrich
NanoFabTx PLA-nano, for synthesis of 100 and 200 nm particles
Sigma-Aldrich
NanoFabTx materials screening kit, for synthesis of polymeric nanoparticles
Sigma-Aldrich
NanoFabTx PEGylated nanoparticle formulation screening kit