Merck
  • Identification of BLNK and BTK as mediators of rituximab-induced programmed cell death by CRISPR screens in GCB-subtype diffuse large B-cell lymphoma.
Identification of BLNK and BTK as mediators of rituximab-induced programmed cell death by CRISPR screens in GCB-subtype diffuse large B-cell lymphoma.
Molecular oncology (2020-06-26)
Emil Aagaard Thomsen, Anne Bruun Rovsing, Mads Valdemar Anderson, Hanne Due, Jinrong Huang, Yonglun Luo, Karen Dybkaer, Jacob Giehm Mikkelsen
ABSTRACT
Diffuse large B-cell lymphoma (DLBCL) is characterized by extensive genetic heterogeneity, and this results in unpredictable responses to the current treatment, R-CHOP, which consists of a cancer drug combination supplemented with the humanized CD20-targeting monoclonal antibody rituximab. Despite improvements in the patient response rate through rituximab addition to the treatment plan, up to 40% of DLBCL patients end in a relapsed or refractory state due to inherent or acquired resistance to the regimen. Here, we employ a lentiviral genome-wide clustered regularly interspaced short palindromic repeats library screening approach to identify genes involved in facilitating the rituximab response in cancerous B cells. Along with the CD20-encoding MS4A1 gene, we identify genes related to B-cell receptor (BCR) signaling as mediators of the intracellular signaling response to rituximab. More specifically, the B-cell linker protein (BLNK) and Bruton's tyrosine kinase (BTK) genes stand out as pivotal genes in facilitating direct rituximab-induced apoptosis through mechanisms that occur alongside complement-dependent cytotoxicity (CDC). Our findings demonstrate that rituximab triggers BCR signaling in a BLNK- and BTK-dependent manner and support the existing notion that intertwined CD20 and BCR signaling pathways in germinal center B-cell-like-subtype DLBCL lead to programmed cell death.
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)