Merck
  • Organic osmolytes increase expression of specific tight junction proteins in skin and alter barrier function in keratinocytes.
Organic osmolytes increase expression of specific tight junction proteins in skin and alter barrier function in keratinocytes.
The British journal of dermatology (2020-04-30)
C El-Chami, A R Foster, C Johnson, R P Clausen, P Cornwell, I S Haslam, M C Steward, R E B Watson, H S Young, C A O'Neill
ABSTRACT
The epidermal barrier is important for water conservation, failure of which is evident in dry-skin conditions. Barrier function is fulfilled by the stratum corneum, tight junctions (TJs, which control extracellular water) and keratinocyte mechanisms, such as organic osmolyte transport, which regulate intracellular water homeostasis. Organic osmolyte transport by keratinocytes is largely unexplored and nothing is known regarding how cellular and extracellular mechanisms of water conservation may interact. We aimed to characterize osmolyte transporters in skin and keratinocytes, and, using transporter inhibitors, to investigate whether osmolytes can modify TJs. Such modification would suggest a possible link between intracellular and extracellular mechanisms of water regulation in skin. Immunostaining and quantitative polymerase chain reaction of organic osmolyte-treated organ-cultured skin were used to identify changes to organic osmolyte transporters, and TJ protein and gene expression. TJ functional assays were performed on organic osmolyte-treated primary human keratinocytes in culture. Immunostaining demonstrated the expression of transporters for betaine, taurine and myo-inositol in transporter-specific patterns. Treatment of human skin with either betaine or taurine increased the expression of claudin-1, claudin-4 and occludin. Osmolyte transporter inhibition abolished this response. Betaine and taurine increased TJ function in primary human keratinocytes in vitro. Treatment of skin with organic osmolytes modulates TJ structure and function, which could contribute to the epidermal barrier. This emphasizes a role for organic osmolytes beyond the maintenance of intracellular osmolarity. This could be harnessed to enhance topical therapies for diseases characterized by skin barrier dysfunction.