Merck
  • Conformationally constrained diketopimelic acid analogues as inhibitors of dihydrodipicolinate synthase.
Conformationally constrained diketopimelic acid analogues as inhibitors of dihydrodipicolinate synthase.
Bioorganic & medicinal chemistry letters (2007-12-14)
Berin A Boughton, Renwick C J Dobson, Juliet A Gerrard, Craig A Hutton
ABSTRACT
Dihydrodipicolinate synthase (DHDPS) is a key enzyme in lysine biosynthesis and a potential antibiotic target. The enzyme catalyses the condensation of (S)-aspartate semi-aldehyde (ASA) and pyruvate to form dihydrodipicolinate. Constrained diketopimelic acid derivatives have been designed as mimics of the acyclic enzyme-bound condensation product of ASA and pyruvate. Several of the compounds are shown to be active, slow-binding inhibitors with improved inhibition of DHDPS.
Product Description
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%
Pricing and availability is not currently available.