Merck
  • Age-induced accumulation of methylmalonic acid promotes tumour progression.
Age-induced accumulation of methylmalonic acid promotes tumour progression.
Nature (2020-08-21)
Ana P Gomes, Didem Ilter, Vivien Low, Jennifer E Endress, Juan Fernández-García, Adam Rosenzweig, Tanya Schild, Dorien Broekaert, Adnan Ahmed, Melanie Planque, Ilaria Elia, Julie Han, Charles Kinzig, Edouard Mullarky, Anders P Mutvei, John Asara, Rafael de Cabo, Lewis C Cantley, Noah Dephoure, Sarah-Maria Fendt, John Blenis
ABSTRACT
The risk of cancer and associated mortality increases substantially in humans from the age of 65 years onwards1-6. Nonetheless, our understanding of the complex relationship between age and cancer is still in its infancy2,3,7,8. For decades, this link has largely been attributed to increased exposure time to mutagens in older individuals. However, this view does not account for the established role of diet, exercise and small molecules that target the pace of metabolic ageing9-12. Here we show that metabolic alterations that occur with age can produce a systemic environment that favours the progression and aggressiveness of tumours. Specifically, we show that methylmalonic acid (MMA), a by-product of propionate metabolism, is upregulated in the serum of older people and functions as a mediator of tumour progression. We traced this to the ability of MMA to induce SOX4 expression and consequently to elicit transcriptional reprogramming that can endow cancer cells with aggressive properties. Thus, the accumulation of MMA represents a link between ageing and cancer progression, suggesting that MMA is a promising therapeutic target for advanced carcinomas.
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Methylmalonic acid, 99%
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)