Merck
Exploring the role of survivin in neuroendocrine neoplasms.
Oncotarget (2020-06-25)
Ahmad Hanif, Sunyoung Lee, Medhavi Gupta, Ankush Chander, Eric D Kannisto, Achamaporn Punnanitinont, Robert Fenstermaker, Michael Ciesielski, Kristopher Attwood, Jingxin Qiu, Sai Yendamuri, Renuka Iyer
ABSTRACT
Neuroendocrine tumors (NETs) are a heterogenous group of tumors. While most NETs have excellent prognosis, certain subsets have aggressive biology and have limited treatment options. We explored the role of survivin in NET as a prognostic and potentially therapeutic marker. Tissue microarrays of 132 patients were stained for survivin using immunohistochemistry (IHC) and correlated with outcomes. Using genomic database, we then correlated survivin (BIRC5) mRNA expression with radiosensitivity index (RSI) in 52 samples of NET. Finally, we studied the effect of radiation on survivin expression in human cell lines and the impact of knock-down of BIRC5 on cell proliferation and radiation sensitivity. We found that survivin positivity by IHC correlated with a shorter survival (overall survival 8.5 years vs. 18.3 years, p < 0.001). There was a positive correlation between BIRC5 expression and RSI (r = 0.234, p < 0.0001). Radiation exposure increased BIRC5 gene expression in a human carcinoid cell line. Knockout of BIRC5 using siRNA reduced proliferation of neuroendocrine cells but did not increase radiation sensitivity. We conclude that survivin expression in NET correlates with an inferior survival and survivin expression in human carcinoid cell lines increases after exposure to ionizing radiation.