Merck
Golgi-Cox Staining Step by Step.
Frontiers in neuroanatomy (2016-04-12)
Sami Zaqout, Angela M Kaindl
ABSTRACT
Golgi staining remains a key method to study neuronal morphology in vivo. Since most protocols delineating modifications of the original staining method lack details on critical steps, establishing this method in a laboratory can be time-consuming and frustrating. Here, we describe the Golgi-Cox staining in such detail that should turn the staining into an easily feasible method for all scientists working in the neuroscience field.
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Pricing and availability is not currently available.
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Pricing and availability is not currently available.