Merck
  • Purification and characterization of transcribed RNAs using gel filtration chromatography.
Purification and characterization of transcribed RNAs using gel filtration chromatography.
Nature protocols (2007-12-15)
Sean A McKenna, Insil Kim, Elisabetta Viani Puglisi, Darrin A Lindhout, Colin Echeverría Aitken, R Andrew Marshall, Joseph D Puglisi
ABSTRACT
RNA synthesis using in vitro transcription by phage T7 RNA polymerase allows preparation of milligram quantities of RNA for biochemical, biophysical and structural investigations. Previous purification approaches relied on gel electrophoretic or gravity-flow chromatography methods. We present here a protocol for the in vitro transcription of RNAs and subsequent purification using fast-performance liquid chromatography. This protocol greatly facilitates production of RNA in a single day from transcription to purification.
Sigma-Aldrich
Cytidine 5′-triphosphate disodium salt, ≥95%