Merck
  • Novel and Efficient Protocol for DNA Coating-Based Identification of DNA-Protein Interaction by Antibody-Mediated Immunodetection.
Novel and Efficient Protocol for DNA Coating-Based Identification of DNA-Protein Interaction by Antibody-Mediated Immunodetection.
Reports of biochemistry & molecular biology (2021-03-03)
Chellathurai Vasantha Niranjan, Selvan Johnson Retnaraj Samue, Venkatachalam Saravanakumar, Selvan Jackson Durairaj
ABSTRACT
Studying protein-protein and protein-DNA interactions are prerequisites for the identification of function and mechanistic role of various proteins in the cell. Protocols for analyzing DNA-based Protein-Protein and Protein-DNA interactions are complicated and need to be simplified for efficient tracking of binding capabilities of various proteins to specific DNA molecules. Here, we demonstrated a simple yet efficient protocol for the identification of DNA coating-based Protein-DNA interaction using antibodymediated immunodetection. Briefly, we have coated specific DNA in the microtiter plate followed by incubating with protein lysate. Specific protein-DNA and/or protein-protein bind with DNA interactions are identified using specific fluorophore-conjugated antibodies. Antibodies are used to detect a protein that is bound to the DNA. Fluorescent-based detection identifies the specific interaction between Protein-DNA with respect to coated DNA fragments. The protocol uses indirect conjugated antibodies and hence the technique is sensitive for effective identification of Protein-DNA interactions. Based on the results we conclude that the demonstrated protocol is simple, efficient and sensitive for identification of Protein-DNA interactions.