Merck
  • Genetically encoded Ca2+ -sensor reveals details of porcine endothelial cell activation upon contact with human serum.
Genetically encoded Ca2+ -sensor reveals details of porcine endothelial cell activation upon contact with human serum.
Xenotransplantation (2020-02-15)
Annegret Wuensch, Petra Kameritsch, Riccardo Sfriso, Eva-Maria Jemiller, Andrea Bähr, Mayuko Kurome, Barbara Kessler, Elisabeth Kemter, Christian Kupatt, Bruno Reichart, Robert Rieben, Eckhard Wolf, Nikolai Klymiuk
ABSTRACT
The activation of the endothelial surface in xenografts is still a poorly understood process and the consequences are unpredictable. The role of Ca2+ -messaging during the activation of endothelial cells is well recognized and routinely measured by synthetic Ca2+ -sensitive fluorophors. However, these compounds require fresh loading immediately before each experiment and in particular when grown in state-of-the-art 3D cell culture systems, endothelial cells are difficult to access with such sensors. Therefore, we developed transgenic pigs expressing a Ca2+ -sensitive protein and examined its principal characteristics. Primary transgenic endothelial cells stimulated by ATP showed a definite and short influx of Ca2+ into the cytosol, whereas exposure to human serum resulted in a more intense and sustained response. Surprisingly, not all endothelial cells reacted identically to a stimulus, rather activation took place in adjacent cells in a timely decelerated way and with distinct intensities. This effect was again more pronounced when cells were stimulated with human serum. Finally, we show clear evidence that antibody binding alone significantly activated endothelial cells, whereas antibody depletion dramatically reduced the stimulatory potential of serum. Transgenic porcine endothelial cells expressing a Ca2+ -sensor represent an interesting tool to dissect factors inducing activation of porcine endothelial cells after exposure to human blood or serum.