Merck
  • Substituted 6-phenyl-2-naphthols. Potent and selective nonsteroidal inhibitors of 17beta-hydroxysteroid dehydrogenase type 1 (17beta-HSD1): design, synthesis, biological evaluation, and pharmacokinetics.
Substituted 6-phenyl-2-naphthols. Potent and selective nonsteroidal inhibitors of 17beta-hydroxysteroid dehydrogenase type 1 (17beta-HSD1): design, synthesis, biological evaluation, and pharmacokinetics.
Journal of medicinal chemistry (2008-07-18)
Sandrine Marchais-Oberwinkler, Patricia Kruchten, Martin Frotscher, Erika Ziegler, Alexander Neugebauer, Umadevi Bhoga, Emmanuel Bey, Ursula Müller-Vieira, Josef Messinger, Hubert Thole, Rolf W Hartmann
ABSTRACT
17beta-Estradiol (E2) is implicated in the genesis and the development of estrogen-dependent diseases. Its concentration is mainly regulated by 17beta-hydroxysteroid dehydrogenase type 1 (17beta-HSD1), which catalyzes the reduction of the weak estrogen estrone (E1) to the highly potent E2. This enzyme is thus an important target for the treatment of hormone-dependent diseases. Thirty-seven novel substituted 6-phenyl-2-naphthols were synthesized and evaluated for 17beta-HSD1 inhibition, selectivity toward 17beta-HSD2 and the estrogen receptors (ERs) alpha and beta, and pharmacokinetic properties. SAR studies revealed that the compounds most likely bind according to binding mode B to the active site, i.e., the 6-phenyl moiety mimicking the steroidal A-ring. While substitution at the phenyl ring decreased activity, introduction of substituents at the naphthol moiety led to highly active compounds, especially in position 1. The 1-phenyl compound 32 showed a very high inhibitory activity for 17beta-HSD1 (IC50 = 20 nM) and good selectivity (17beta-HSD2 and ERs) and pharmacokinetic properties after peroral application.
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications