Merck
  • miR-196b-5p inhibits proliferation of Wharton's jelly umbilical cord stem cells.
miR-196b-5p inhibits proliferation of Wharton's jelly umbilical cord stem cells.
FEBS open bio (2020-11-19)
Xiao Han, Haoqing Yang, Huina Liu, Chen Zhang, Yangyang Cao, Zhipeng Fan, Ruitang Shi
ABSTRACT
Human umbilical cord mesenchymal stem cells can be obtained from different parts of the umbilical cord, including Wharton's jelly. Transplantation of Wharton's jelly umbilical cord stem cells (WJCMSCs) is a promising strategy for the treatment of various diseases. However, the molecular mechanisms underlying the proliferation of WJCMSCs are incompletely understood. Here, we report that overexpression of miR-196b-5p in WJCMSCs suppresses proliferation and arrests the cell cycle in G0/G1 phase, whereas knockdown of miR-196b-5p promotes WJCMSC proliferation and cell-cycle progression. Moreover, miR-196b-5p overexpression resulted in decreased levels of Cyclin A, Cyclin D, Cyclin E and cyclin-dependent kinases 2 and increased levels of p15INK4b , whereas miR-196b-5p knockdown had the opposite effects. In conclusion, our data suggests that miR-196b-5p inhibits WJCMSC proliferation by enhancing G0/G1-phase arrest.
Sigma-Aldrich
Anti-PLCγ-1 Antibody, Upstate®, from mouse
Sigma-Aldrich
Anti-Cdk4 Antibody, clone DCS-35, clone DCS-35, Chemicon®, from mouse
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Cyclin E Antibody, clone HE12, clone HE12, Upstate®, from mouse