Merck
  • SENP1-Sirt3 Signaling Controls Mitochondrial Protein Acetylation and Metabolism.
SENP1-Sirt3 Signaling Controls Mitochondrial Protein Acetylation and Metabolism.
Molecular cell (2019-07-16)
Tianshi Wang, Ying Cao, Quan Zheng, Jun Tu, Wei Zhou, Jianli He, Jie Zhong, Yalan Chen, Jiqiu Wang, Rong Cai, Yong Zuo, Bo Wei, Qiuju Fan, Jie Yang, Yicheng Wu, Jing Yi, Dali Li, Mingyao Liu, Chuangui Wang, Aiwu Zhou, Yu Li, Xuefeng Wu, Wen Yang, Y Eugene Chin, Guoqiang Chen, Jinke Cheng
ABSTRACT
Sirt3, as a major mitochondrial nicotinamide adenine dinucleotide (NAD)-dependent deacetylase, is required for mitochondrial metabolic adaption to various stresses. However, how to regulate Sirt3 activity responding to metabolic stress remains largely unknown. Here, we report Sirt3 as a SUMOylated protein in mitochondria. SUMOylation suppresses Sirt3 catalytic activity. SUMOylation-deficient Sirt3 shows elevated deacetylation on mitochondrial proteins and increased fatty acid oxidation. During fasting, SUMO-specific protease SENP1 is accumulated in mitochondria and quickly de-SUMOylates and activates Sirt3. SENP1 deficiency results in hyper-SUMOylation of Sirt3 and hyper-acetylation of mitochondrial proteins, which reduces mitochondrial metabolic adaption responding to fasting. Furthermore, we find that fasting induces SENP1 translocation into mitochondria to activate Sirt3. The studies on mice show that Sirt3 SUMOylation mutation reduces fat mass and antagonizes high-fat diet (HFD)-induced obesity via increasing oxidative phosphorylation and energy expenditure. Our results reveal that SENP1-Sirt3 signaling modulates Sirt3 activation and mitochondrial metabolism during metabolic stress.
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Roche
Endoproteinase Glu-C (V8 Protease), from Staphylococcus aureus V8
Sigma-Aldrich
Palmitoyl-L-carnitine chloride, ≥98% (TLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)