Merck
  • PD-L1-mediated gasdermin C expression switches apoptosis to pyroptosis in cancer cells and facilitates tumour necrosis.
PD-L1-mediated gasdermin C expression switches apoptosis to pyroptosis in cancer cells and facilitates tumour necrosis.
Nature cell biology (2020-09-16)
Junwei Hou, Rongce Zhao, Weiya Xia, Chiung-Wen Chang, Yun You, Jung-Mao Hsu, Lei Nie, Yeh Chen, Yu-Chuan Wang, Chunxiao Liu, Wei-Jan Wang, Yun Wu, Baozhen Ke, Jennifer L Hsu, Kebin Huang, Zu Ye, Yi Yang, Xianghou Xia, Yintao Li, Chia-Wei Li, Bin Shao, John A Tainer, Mien-Chie Hung
ABSTRACT
Although pyroptosis is critical for macrophages against pathogen infection, its role and mechanism in cancer cells remains unclear. PD-L1 has been detected in the nucleus, with unknown function. Here we show that PD-L1 switches TNFα-induced apoptosis to pyroptosis in cancer cells, resulting in tumour necrosis. Under hypoxia, p-Stat3 physically interacts with PD-L1 and facilitates its nuclear translocation, enhancing the transcription of the gasdermin C (GSDMC) gene. GSDMC is specifically cleaved by caspase-8 with TNFα treatment, generating a GSDMC N-terminal domain that forms pores on the cell membrane and induces pyroptosis. Nuclear PD-L1, caspase-8 and GSDMC are required for macrophage-derived TNFα-induced tumour necrosis in vivo. Moreover, high expression of GSDMC correlates with poor survival. Antibiotic chemotherapy drugs induce pyroptosis in breast cancer. These findings identify a non-immune checkpoint function of PD-L1 and provide an unexpected concept that GSDMC/caspase-8 mediates a non-canonical pyroptosis pathway in cancer cells, causing tumour necrosis.
Sigma-Aldrich
Z-Ile-Glu(O-ME)-Thr-Asp(O-Me) fluoromethyl ketone, ≥90% (TLC), powder
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Supelco
Busulfan, analytical standard, for drug analysis
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Thio-TEPA
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Terbium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Terbium(III) chloride, anhydrous, powder, 99.9% trace metals basis
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile
USP
Topotecan hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carmustine, ≥98%
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
Chlorambucil
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Epirubicin hydrochloride, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Dacarbazine, British Pharmacopoeia (BP) Reference Standard
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)