Merck
  • Cathepsin A contributes to left ventricular remodeling by degrading extracellular superoxide dismutase in mice.
Cathepsin A contributes to left ventricular remodeling by degrading extracellular superoxide dismutase in mice.
The Journal of biological chemistry (2020-07-11)
Mathias Hohl, Manuel Mayr, Lisa Lang, Alexander G Nickel, Javier Barallobre-Barreiro, Xiaoke Yin, Thimoteus Speer, Simina-Ramona Selejan, Claudia Goettsch, Katharina Erb, Claudia Fecher-Trost, Jan-Christian Reil, Benedikt Linz, Sven Ruf, Thomas Hübschle, Christoph Maack, Michael Böhm, Thorsten Sadowski, Dominik Linz, Mathias Hohl, Manuel Mayr, Lisa Lang, Alexander G Nickel, Javier Barallobre-Barreiro, Xiaoke Yin, Thimoteus Speer, Simina-Ramona Selejan, Claudia Goettsch, Katharina Erb, Claudia Fecher-Trost, Jan-Christian Reil, Benedikt Linz, Sven Ruf, Thomas Hübschle, Christoph Maack, Michael Böhm, Thorsten Sadowski, Dominik Linz
ABSTRACT
In the heart, the serine carboxypeptidase cathepsin A (CatA) is distributed between lysosomes and the extracellular matrix (ECM). CatA-mediated degradation of extracellular peptides may contribute to ECM remodeling and left ventricular (LV) dysfunction. Here, we aimed to evaluate the effects of CatA overexpression on LV remodeling. A proteomic analysis of the secretome of adult mouse cardiac fibroblasts upon digestion by CatA identified the extracellular antioxidant enzyme superoxide dismutase (EC-SOD) as a novel substrate of CatA, which decreased EC-SOD abundance 5-fold. In vitro, both cardiomyocytes and cardiac fibroblasts expressed and secreted CatA protein, and only cardiac fibroblasts expressed and secreted EC-SOD protein. Cardiomyocyte-specific CatA overexpression and increased CatA activity in the LV of transgenic mice (CatA-TG) reduced EC-SOD protein levels by 43%. Loss of EC-SOD-mediated antioxidative activity resulted in significant accumulation of superoxide radicals (WT, 4.54 μmol/mg tissue/min; CatA-TG, 8.62 μmol/mg tissue/min), increased inflammation, myocyte hypertrophy (WT, 19.8 μm; CatA-TG, 21.9 μm), cellular apoptosis, and elevated mRNA expression of hypertrophy-related and profibrotic marker genes, without affecting intracellular detoxifying proteins. In CatA-TG mice, LV interstitial fibrosis formation was enhanced by 19%, and the type I/type III collagen ratio was shifted toward higher abundance of collagen I fibers. Cardiac remodeling in CatA-TG was accompanied by an increased LV weight/body weight ratio and LV end diastolic volume (WT, 50.8 μl; CatA-TG, 61.9 μl). In conclusion, CatA-mediated EC-SOD reduction in the heart contributes to increased oxidative stress, myocyte hypertrophy, ECM remodeling, and inflammation, implicating CatA as a potential therapeutic target to prevent ventricular remodeling.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution